Orders LinkContactUsLinkComponent
Meker burners
Meker burners
Meker burner, with head Ø 43 mm, LPG, 43 mm
Katalog # RLAA4101/51
Leverandør:   R L ENTERPRISES
Meker burners
Katalog # RLAA4101/51
Leverandør:  R L ENTERPRISES
Meker burners
Meker burner, with head Ø 43 mm, LPG, 43 mm
Price
3.650,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Nickel plated brass. The top of the burner is fitted with a perforated steel head, which gives multiple small but very hot flames. It is not fitted with an air regulator. Burners have diecast base painted in a blue chemically resistant acrylic finish.
Specifikationer
  • Beskrivelse:
    Meker burner, with head Ø 43 mm
  • Seq:
    07
  • Ø hoved:
    43 mm
  • Til:
    LPG