Meker burners
Meker burner, with head Ø 43 mm, LPG, 43 mm
Katalog # RLAA4101/51
Leverandør: R L ENTERPRISES
Gå til:
Product Details & Documents
Nickel plated brass. The top of the burner is fitted with a perforated steel head, which gives multiple small but very hot flames. It is not fitted with an air regulator. Burners have diecast base painted in a blue chemically resistant acrylic finish.
Specifikationer
- Beskrivelse:Meker burner, with head Ø 43 mm
- Seq:07
- Ø hoved:43 mm
- Til:LPG