Orders LinkContactUsLinkComponent
Meker burners
Meker burners
Meker burner, with head Ø 30 mm, Coal, 30 mm
Katalog # RLAA4101/4
Leverandør:   R L ENTERPRISES
Meker burners
Katalog # RLAA4101/4
Leverandør:  R L ENTERPRISES
Meker burners
Meker burner, with head Ø 30 mm, Coal, 30 mm
Price
1.380,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Nickel plated brass. The top of the burner is fitted with a perforated steel head, which gives multiple small but very hot flames. It is not fitted with an air regulator. Burners have diecast base painted in a blue chemically resistant acrylic finish.
Specifikationer
  • Beskrivelse:
    Meker burner, with head Ø 30 mm
  • Seq:
    06
  • Ø hoved:
    30 mm
  • Til:
    Coal