Meker burners
Meker burner, with head Ø 24 mm, Coal, 24 mm
Katalog # RLAA4101/3
Leverandør: R L ENTERPRISES
Gå til:
Product Details & Documents
Nickel plated brass. The top of the burner is fitted with a perforated steel head, which gives multiple small but very hot flames. It is not fitted with an air regulator. Burners have diecast base painted in a blue chemically resistant acrylic finish.
Specifikationer
- Beskrivelse:Meker burner, with head Ø 24 mm
- Seq:04
- Ø hoved:24 mm
- Til:Coal