Meker burners
Meker burner, with head Ø 20 mm, LPG, 20 mm
RLAA4101/21
Nickel plated brass. The top of the burner is fitted with a perforated steel head, which gives multiple small but very hot flames. It is not fitted with an air regulator. Burners have diecast base painted in a blue chemically resistant acrylic finish.
- Beskrivelse:Meker burner, with head Ø 20 mm
- Seq:01
- Ø hoved:20 mm
- Til:LPG