Orders LinkContactUsLinkComponent
Meker burners
Meker burner, with head Ø 20 mm, LPG, 20 mm
  RLAA4101/21
Meker burners
  RLAA4101/21
Meker burners
Meker burner, with head Ø 20 mm, LPG, 20 mm
922,00 kr
1 * 1 Items
Nickel plated brass. The top of the burner is fitted with a perforated steel head, which gives multiple small but very hot flames. It is not fitted with an air regulator. Burners have diecast base painted in a blue chemically resistant acrylic finish.
  • Beskrivelse:
    Meker burner, with head Ø 20 mm
  • Seq:
    01
  • Ø hoved:
    20 mm
  • Til:
    LPG