Orders LinkContactUsLinkComponent
Retord rods, M 10 thread
Retord rods, M 10 thread
12,5 mm, 1000 mm
  241-0194
Retord rods, M 10 thread
  241-0194
Retord rods, M 10 thread
Retord rods, M 10 thread
12,5 mm, 1000 mm
240,00 kr
1 * 1 Items
Retort rods with M10 thread, made of zinc plated steel.
  • Ø:
    12,5 mm
  • Længde:
    1000 mm
  • Seq:
    04