Orders LinkContactUsLinkComponent
Retord rods, M 10 thread
12,5 mm, 600 mm
  241-0189
Retord rods, M 10 thread
  241-0189
Retord rods, M 10 thread
Retord rods, M 10 thread
12,5 mm, 600 mm
183,00 kr
1 * 1 Items
Retort rods with M10 thread, made of zinc plated steel.
  • Ø:
    12,5 mm
  • Længde:
    600 mm
  • Seq:
    02