Retord rods, M 10 thread
10 mm, 500 mm
Catalog # 241-0186
Supplier: R L ENTERPRISES
Jump to:
Product Details & Documents
Retort rods with M10 thread, made of zinc plated steel.
Specifications
- Ø:10 mm
- Længde:500 mm
- Seq:01