Double bosshead
Bosshead with blue plastic headed screw, Støbt legering, 16 mm
Katalog # 241-7867
Leverandør: R L ENTERPRISES
Gå til:
Product Details & Documents
Bossheads made from nickel plated cast alloy.
- Either with or without blue plastic headed nickel plated screws
- For use with rods up to 16 mm
Specifikationer
- Beskrivelse:Bosshead with blue plastic headed screw
- Materiale:Støbt legering
- Til stang Ø:16 mm