Formaldehyd 4% stabilised, buffered (pH 7,0 ±0,1)
Plastic bottle, 1 L
Leverandør: VWR Chemicals
Formaldehyd 4% stabilised, buffered (pH 7,0 ±0,1)
Katalog # 9713.1000
Leverandør: VWR Chemicals
CAS nummer: 50-00-0
Gå til:
Product Details & Documents
(= 10% Formaline solution)
Synonymer: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS nummer:50-00-0
- Formel:CH₂O
- MDL nummer:MFCD00003274
- Molecular Weight:30.03 g/mol
- :stabiliseret med methanol 0,5-1,5 %
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-001-8
Certifikater
Søg efter partinummer10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 25L Plastic drum for liquids DIN 61 (ZPACK13952500)
Specifikationer
Specifikationstestresultater
- Assay (Formaldehyde):3,5 - 4,5 %
- pH (20 °C):6,9 - 7,1
- Methanol:0,5 - 1,5 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:1 L



