Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Natriumhypochlorit (14% Cl₂) in aqueous solution, TECHNICAL

Natriumhypochlorit (14% Cl₂) in aqueous solution, TECHNICAL

Plastic drum, 25 L
Katalog # 90350.9025
Ætsende
Miljø
Leverandør:   VWR Chemicals
Natriumhypochlorit (14% Cl₂) in aqueous solution, TECHNICAL
Katalog # 90350.9025
Leverandør:  VWR Chemicals
CAS nummer:  7681-52-9
Natriumhypochlorit (14% Cl₂) in aqueous solution, TECHNICAL
Natriumhypochlorit (14% Cl₂) in aqueous solution, TECHNICAL

Plastic drum, 25 L

CAS nummer:  7681-52-9
Price
1.490,00 kr
Log ind for tilgængelighed
1 * 25 L
Product Details & Documents

Da produktet ikke er stabiliseret mister det normalt styrke under opbevaring.

  • CAS nummer:7681-52-9
  • Formel:ClNaO
  • MDL nummer:MFCD00011120
  • Molecular Weight:74.44 g/mol
  • Kogende Pt:111 °C (1013 hPa)
  • Smeltende Pt:–30…–20 °C
  • Tæthed:1,22 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-668-3
  • FN:1791
  • ADR:8,II
Specifikationer

Specifikationstestresultater

  • Assay (Active chlorine)(W/V)(at filling):
    Min. 11.5 %
  • Identification:
    Passes test

Specifikationer

  • Pakning:
    Plastic drum
  • Pakn:
    25 L

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.