Polyadenylic acid (poly-A)
Polyadenylic acid (poly-A), 500 mg
Leverandør: Cytiva
Gå til:
Product Details & Documents
Poly(A) (polyadenylic acid) can be used to evaluate the binding of potential therapeutic agents to mRNA in drug development research.
- Average chain length of >200 nucleotides
- Ribonuclease tested
Specifikationer
Specifikationer
- Pakn:500 mg
- Beskrivelse:Polyadenylic acid (poly-A)