Orders LinkContactUsLinkComponent
Polyadenylic acid (poly-A)
Polyadenylic acid (poly-A)
Polyadenylic acid (poly-A), 100 mg
Katalog # 27-4110-01
Leverandør:   Cytiva
Polyadenylic acid (poly-A)
Katalog # 27-4110-01
Leverandør:  Cytiva
CAS nummer:  
Polyadenylic acid (poly-A)
Polyadenylic acid (poly-A)
Polyadenylic acid (poly-A), 100 mg
CAS nummer:  
Price
2.240,00 kr
Log ind for tilgængelighed
1 * 100 mg
Product Details & Documents
Poly(A) (polyadenylic acid) can be used to evaluate the binding of potential therapeutic agents to mRNA in drug development research.
  • Average chain length of >200 nucleotides
  • Ribonuclease tested
Specifikationer
Specifikationer
  • Pakn:
    100 mg
  • Beskrivelse:
    Polyadenylic acid (poly-A)