Polyadenylic acid (poly-A)
Polyadenylic acid (poly-A), 100 mg
Leverandør: Cytiva
Gå til:
Product Details & Documents
Poly(A) (polyadenylic acid) can be used to evaluate the binding of potential therapeutic agents to mRNA in drug development research.
- Average chain length of >200 nucleotides
- Ribonuclease tested
Documents
Certifikater
Søg efter partinummerCytiva CyDye Licensing Statements Cytiva General Licensing Statements
Yderligere produktdetaljer
Specifikationer
Specifikationer
- Pakn:100 mg
- Beskrivelse:Polyadenylic acid (poly-A)