Orders LinkContactUsLinkComponent
Tilbehør og reservedele
Tilbehør og reservedele
Reservedelssæt til membraner/ventiler til MZ 8C/MD 8C/ME 16C
Catalog # 181-1678
Supplier:   VACUUBRAND
Tilbehør og reservedele
Catalog # 181-1678
Supplier:  VACUUBRAND
Tilbehør og reservedele
Reservedelssæt til membraner/ventiler til MZ 8C/MD 8C/ME 16C
Price
8.540,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Versions available for pressure and vacuum fine tuning. Easy to assemble.
Ideal for solvent recovery. Emission condensers allow recovery rates up to almost 100% without costly refrigeration packages or cold traps. The AK + EK combination brings together the advantages of inlet separators, to prevent droplets and particles entering and damaging the pump, and promoting efficient solvent recovery.
Specifications
  • Beskrivelse:
    Reservedelssæt til membraner/ventiler til MZ 8C/MD 8C/ME 16C