Tilbehør og reservedele
Reservedelssæt til membraner/ventiler til MZ 8/MD 8/ME 16
Katalog # 181-1674
Leverandør: VACUUBRAND
Gå til:
Product Details & Documents
Versions available for pressure and vacuum fine tuning. Easy to assemble.
Ideal for solvent recovery. Emission condensers allow recovery rates up to almost 100% without costly refrigeration packages or cold traps. The AK + EK combination brings together the advantages of inlet separators, to prevent droplets and particles entering and damaging the pump, and promoting efficient solvent recovery.
Specifikationer
- Beskrivelse:Reservedelssæt til membraner/ventiler til MZ 8/MD 8/ME 16