Orders LinkContactUsLinkComponent
Tilbehør og reservedele
Tilbehør og reservedele
Reservedelssæt til membraner/ventiler til ME 2C
Katalog # 181-1675
Leverandør:   VACUUBRAND
Tilbehør og reservedele
Katalog # 181-1675
Leverandør:  VACUUBRAND
Tilbehør og reservedele
Reservedelssæt til membraner/ventiler til ME 2C
Price
536,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Versions available for pressure and vacuum fine tuning. Easy to assemble.
Ideal for solvent recovery. Emission condensers allow recovery rates up to almost 100% without costly refrigeration packages or cold traps. The AK + EK combination brings together the advantages of inlet separators, to prevent droplets and particles entering and damaging the pump, and promoting efficient solvent recovery.
Specifikationer
  • Beskrivelse:
    Reservedelssæt til membraner/ventiler til ME 2C