Orders LinkContactUsLinkComponent
Tilbehør og reservedele
Reservedelssæt til membraner/ventiler til ME 2
  181-1671
 :   VACUUBRAND
Tilbehør og reservedele
  181-1671
Tilbehør og reservedele
Reservedelssæt til membraner/ventiler til ME 2
527,00 kr
1 * 1 Items
Versions available for pressure and vacuum fine tuning. Easy to assemble.
Ideal for solvent recovery. Emission condensers allow recovery rates up to almost 100% without costly refrigeration packages or cold traps. The AK + EK combination brings together the advantages of inlet separators, to prevent droplets and particles entering and damaging the pump, and promoting efficient solvent recovery.
  • Beskrivelse:
    Reservedelssæt til membraner/ventiler til ME 2