Accessories
pH electrode, round tip, with sealed combination double junction, Ag/AgCl reference, electrolyte bridge of 4M KCl saturated with Ag/AgCl, outer electrolyte bridge of 1M KNO 3, annular ceramic reference junction and low profile
pH membrane
pH membrane
Supplier: Cytiva
Jump to:
Product Details & Documents
Documents
Specifications
- Beskrivelse:pH electrode, round tip, with sealed combination double junction, Ag/AgCl reference, electrolyte bridge of 4M KCl saturated with Ag/AgCl, outer electrolyte bridge of 1M KNO 3, annular ceramic reference junction and low profile
pH membrane - Cat. No.:18-1111-26
- B×D×H:260×370×100 mm
- pH - Nøjagtighed:± 0,1 pH-enhed, temperaturkompenseret
- Output:Analog, 0 - 1 V, 4 - 20 mA
- pH - Område:0 - 14 (specifikationerne gælder fra 2 - 12)