Reducing cross pieces with small flanges
Cross Piece, DN 25/25/10/10
Catalog # 185-2481
Supplier: VACUUBRAND
Jump to:
Product Details & Documents
Reducing cross pieces with small flanges KF made of aluminium or stainless steel.
Used for vacuum system.
Documents
MB_Messkreuzstuecke_KF
Technical Drawing
Specifications
- Beskrivelse:Cross Piece, DN 25/25/10/10
- Seq:03