Orders LinkContactUsLinkComponent
Cross pieces with small flanges
Cross pieces with small flanges
Cross pieces with small flanges KF, DN 16/16/16/16
Katalog # 185-2462
Leverandør:   VACUUBRAND
Cross pieces with small flanges
Katalog # 185-2462
Leverandør:  VACUUBRAND
Cross pieces with small flanges
Cross pieces with small flanges
Cross pieces with small flanges KF, DN 16/16/16/16
Price
2.250,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Cross pieces with small flanges KF made of aluminium or stainless steel.
Used for vacuum system.
Documents
Specifikationer
  • Beskrivelse:
    Cross pieces with small flanges KF, DN 16/16/16/16
  • Seq:
    06