T-pieces with small flanges
T-pieces with small flanges KFDN 10/10/10
Katalog # 185-2441
Leverandør: VACUUBRAND
Gå til:
Product Details & Documents
T-pieces with small flanges KF made of aluminium or stainless steel.
Used for vacuum system.
Documents
MB_T_Stuecke_KF
Teknisk tegning
Specifikationer
- Beskrivelse:T-pieces with small flanges KFDN 10/10/10
- Seq:05