Orders LinkContactUsLinkComponent
Small flanges KF, screw-in
Small flanges KF, screw-in
Small flanges KF, screw-in, male thread, DN 25, Rustfrit stål
Catalog # VACU20672102
Supplier:   VACUUBRAND
Small flanges KF, screw-in
Catalog # VACU20672102
Supplier:  VACUUBRAND
Small flanges KF, screw-in
Small flanges KF, screw-in, male thread, DN 25, Rustfrit stål
Price
1.330,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Small flanges made of stainless steel with sealing ring made of NBR.
Documents
Specifications
  • Beskrivelse:
    Small flanges KF, screw-in, male thread, DN 25
  • Materiale:
    Rustfrit stål
  • Seq:
    03