Orders LinkContactUsLinkComponent
Small flanges KF, screw-in
Small flanges KF, screw-in, male thread, DN 16, Rustfrit stål
Katalog # VACU20672101
Leverandør:   VACUUBRAND
Small flanges KF, screw-in
Katalog # VACU20672101
Leverandør:  VACUUBRAND
Small flanges KF, screw-in
Small flanges KF, screw-in, male thread, DN 16, Rustfrit stål
Price
1.060,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Small flanges made of stainless steel with sealing ring made of NBR.
Specifikationer
  • Beskrivelse:
    Small flanges KF, screw-in, male thread, DN 16
  • Materiale:
    Rustfrit stål
  • Seq:
    02