Small flanges KF, screw-in
Small flanges KF, screw-in, male thread, DN 10, Rustfrit stål
Katalog # VACU20672100
Leverandør: VACUUBRAND
Gå til:
Product Details & Documents
Small flanges made of stainless steel with sealing ring made of NBR.
Documents
MB_KFEinschraubflansche
Teknisk tegning
Specifikationer
- Beskrivelse:Small flanges KF, screw-in, male thread, DN 10
- Materiale:Rustfrit stål
- Seq:01