Blind small flanges
Blanking flange, DN 40, Rustfrit stål
185-2114
Blind small flanges KF made of aluminium or stainless steel.
Used for vacuum system.
- Beskrivelse:Blanking flange, DN 40
- Materiale:Rustfrit stål
- Seq:09
Blind small flanges KF made of aluminium or stainless steel.
Used for vacuum system.