Blind small flanges
Blanking flange, DN 25, Rustfrit stål
185-2113
Blind small flanges KF made of aluminium or stainless steel.
Used for vacuum system.
- Beskrivelse:Blanking flange, DN 25
- Materiale:Rustfrit stål
- Seq:08
Blind small flanges KF made of aluminium or stainless steel.
Used for vacuum system.