Blind small flanges
Blanking flange, DN 16, Rustfrit stål
Catalog # 185-2112
Supplier: VACUUBRAND
Jump to:
Product Details & Documents
Blind small flanges KF made of aluminium or stainless steel.
Used for vacuum system.
Specifications
- Beskrivelse:Blanking flange, DN 16
- Materiale:Rustfrit stål
- Seq:07