Reducing cross pieces with small flanges
Cross Piece, DN 40/40/16/16
Katalog # 185-2472
Leverandør: VACUUBRAND
Gå til:
Product Details & Documents
Reducing cross pieces with small flanges KF made of aluminium or stainless steel.
Used for vacuum system.
Documents
MB_Messkreuzstuecke_KF
Technical Drawing
Specifikationer
- Beskrivelse:Cross Piece, DN 40/40/16/16
- Seq:02