Cross pieces with small flanges
Cross pieces with small flanges KF, DN 16/16/16/16
Katalog # 185-2452
Leverandør: VACUUBRAND
Gå til:
Product Details & Documents
Cross pieces with small flanges KF made of aluminium or stainless steel.
Used for vacuum system.
Documents
MB_Kreuzstuecke_KF
Technical Drawing
Specifikationer
- Beskrivelse:Cross pieces with small flanges KF, DN 16/16/16/16
- Seq:02