Orders LinkContactUsLinkComponent
T-pieces with small flanges
T-pieces with small flanges
T-pieces with small flanges KFDN 25/25/25
Katalog # 185-2433
Leverandør:   VACUUBRAND
T-pieces with small flanges
Katalog # 185-2433
Leverandør:  VACUUBRAND
T-pieces with small flanges
T-pieces with small flanges KFDN 25/25/25
Price
740,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
T-pieces with small flanges KF made of aluminium or stainless steel.
Used for vacuum system.
Documents
Specifikationer
  • Beskrivelse:
    T-pieces with small flanges KFDN 25/25/25
  • Seq:
    03