Orders LinkContactUsLinkComponent
Small flanges with weld stub long
Small flanges with weld stub long
Small flange, KF, DN 25
Katalog # VACU20662105
Leverandør:   VACUUBRAND
Small flanges with weld stub long
Katalog # VACU20662105
Leverandør:  VACUUBRAND
Small flanges with weld stub long
Small flanges with weld stub long
Small flange, KF, DN 25
Price
797,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These flanges are available with pipe socket.
  • Made of stainless steel
  • Used for welding
Documents
Specifikationer
  • Beskrivelse:
    Small flange, KF, DN 25
  • Seq:
    04