Small flanges with weld stub long
Small flange, KF, DN 25
Katalog # VACU20662105
Leverandør: VACUUBRAND
Gå til:
Product Details & Documents
These flanges are available with pipe socket.
- Made of stainless steel
- Used for welding
Documents
MB_Kleinflansche_Rohransatz_lang
Teknisk tegning
Specifikationer
- Beskrivelse:Small flange, KF, DN 25
- Seq:04