Small flanges with weld stub long
Small flange, KF, DN 16
Catalog # VACU20662104
Supplier: VACUUBRAND
Jump to:
Product Details & Documents
These flanges are available with pipe socket.
- Made of stainless steel
- Used for welding
Documents
MB_Kleinflansche_Rohransatz_lang
Technical Drawing
Specifications
- Beskrivelse:Small flange, KF, DN 16
- Seq:02