Orders LinkContactUsLinkComponent
Small flanges with weld stub long
Small flanges with weld stub long
Small flange, KF, DN 16
Catalog # VACU20662104
Supplier:   VACUUBRAND
Small flanges with weld stub long
Catalog # VACU20662104
Supplier:  VACUUBRAND
Small flanges with weld stub long
Small flanges with weld stub long
Small flange, KF, DN 16
Price
611,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These flanges are available with pipe socket.
  • Made of stainless steel
  • Used for welding
Documents
Specifications
  • Beskrivelse:
    Small flange, KF, DN 16
  • Seq:
    02