Small flanges with weld stub long
Small flange, KF, DN 20
Katalog # VACU20662101
Leverandør: VACUUBRAND
Gå til:
Product Details & Documents
These flanges are available with pipe socket.
- Made of stainless steel
- Used for welding
Specifikationer
- Beskrivelse:Small flange, KF, DN 20
- Seq:03
- Cat. No.:VACU20662101