Orders LinkContactUsLinkComponent
Small flanges with weld stub long
Small flange, KF, DN 20
Katalog # VACU20662101
Leverandør:   VACUUBRAND
Small flanges with weld stub long
Katalog # VACU20662101
Leverandør:  VACUUBRAND
Small flanges with weld stub long
Small flanges with weld stub long
Small flange, KF, DN 20
Price
753,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These flanges are available with pipe socket.
  • Made of stainless steel
  • Used for welding
Specifikationer
  • Beskrivelse:
    Small flange, KF, DN 20
  • Seq:
    03
  • Cat. No.:
    VACU20662101