Mohr pinch cocks
70 mm
Katalog # 229-0334
Leverandør: USBECK Laborgeräte
Gå til:
Product Details & Documents
Nickel-plated brass. For regulating or stopping the flow of liquids in tube connections.
- Very good thermal resistance
- Easy handling
- Easy cleaning
Specifikationer
- Længde:70 mm