Mohr pinch cocks
60 mm
Catalog # 229-0333
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
Nickel-plated brass. For regulating or stopping the flow of liquids in tube connections.
- Very good thermal resistance
- Easy handling
- Easy cleaning
Specifications
- Længde:60 mm