Orders LinkContactUsLinkComponent
Mohr pinch cocks
Mohr pinch cocks
40 mm
Katalog # 229-0331
Leverandør:   USBECK Laborgeräte
Mohr pinch cocks
Katalog # 229-0331
Leverandør:  USBECK Laborgeräte
Mohr pinch cocks
Mohr pinch cocks
40 mm
Price
63,30 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Nickel-plated brass. For regulating or stopping the flow of liquids in tube connections.
  • Very good thermal resistance
  • Easy handling
  • Easy cleaning
Specifikationer
  • Længde:
    40 mm