Orders LinkContactUsLinkComponent
Diamond pen
Diamond pen
Diamantpen, kuglepen, 140 mm
Katalog # 201-0392
Leverandør:   USBECK Laborgeräte
Diamond pen
Katalog # 201-0392
Leverandør:  USBECK Laborgeräte
Diamond pen
Diamond pen
Diamantpen, kuglepen, 140 mm
Price
403,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Diamond tip in metal holder.
  • Available with wooden handle or in the form of a ballpoint pen
  • Not suitable for engraving
  • 140 mm long
For labelling glass and metal surfaces.
Specifikationer
  • Beskrivelse:
    Diamantpen, kuglepen, 140 mm