Orders LinkContactUsLinkComponent
Diamond pen
Diamantpen, træskaft
  201-0391
Diamond pen
  201-0391
Diamond pen
Diamond pen
Diamantpen, træskaft
389,00 kr
1 * 1 Items
Diamond tip in metal holder.
  • Available with wooden handle or in the form of a ballpoint pen
  • Not suitable for engraving
  • 140 mm long
For labelling glass and metal surfaces.
  • Beskrivelse:
    Diamantpen, træskaft