Glass cutter
Udskiftningsmetalblad
Catalog # 201-9011
Supplier: USBECK Laborgeräte
Jump to:
Product Details & Documents
Plastic handle with hardened metal blade.
- Clean and precise cutting
- Replaceable hardened metal blades
- Easy to change blade
Length 185 mm, blade dimensions 40×20 mm.
Specifications
- Beskrivelse:Udskiftningsmetalblad