Orders LinkContactUsLinkComponent
Glass cutter
Glass cutter
Udskiftningsmetalblad
Catalog # 201-9011
Supplier:   USBECK Laborgeräte
Glass cutter
Catalog # 201-9011
Glass cutter
Glass cutter
Udskiftningsmetalblad
Price
269,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Plastic handle with hardened metal blade.
  • Clean and precise cutting
  • Replaceable hardened metal blades
  • Easy to change blade
Length 185 mm, blade dimensions 40×20 mm.
Specifications
  • Beskrivelse:
    Udskiftningsmetalblad