Orders LinkContactUsLinkComponent
Spoon spatulas, broad shape
Spoon spatulas, broad shape
210 mm, 30 mm
Katalog # USBE3334
Leverandør:   USBECK Laborgeräte
Spoon spatulas, broad shape
Katalog # USBE3334
Leverandør:  USBECK Laborgeräte
Spoon spatulas, broad shape
Spoon spatulas, broad shape
210 mm, 30 mm
Price
59,70 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Rigid spoon spatulas made of stainless steel. With spoon end and one flat end.
Specifikationer
  • Længde:
    210 mm
  • Skestr.:
    30 mm