Orders LinkContactUsLinkComponent
Spoon spatulas, broad shape
Spoon spatulas, broad shape
150 mm, 28 mm
Katalog # USBE3332
Leverandør:   USBECK Laborgeräte
Spoon spatulas, broad shape
Katalog # USBE3332
Leverandør:  USBECK Laborgeräte
Spoon spatulas, broad shape
Spoon spatulas, broad shape
150 mm, 28 mm
Price
46,60 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Rigid spoon spatulas made of stainless steel. With spoon end and one flat end.
Specifikationer
  • Længde:
    150 mm
  • Skestr.:
    28 mm