Orders LinkContactUsLinkComponent
Double spatulas for mortars
Double spatulas for mortars
210 mm, 20 mm
Katalog # 231-0447
Leverandør:   USBECK Laborgeräte
Double spatulas for mortars
Katalog # 231-0447
Leverandør:  USBECK Laborgeräte
Double spatulas for mortars
Double spatulas for mortars
210 mm, 20 mm
Price
53,80 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel.
  • Rigid
  • With two ends
Specifikationer
  • Længde:
    210 mm
  • Bladbredde:
    20 mm