Orders LinkContactUsLinkComponent
Clamps
Clamps
30 - 80 mm
Catalog # 241-7547
Supplier:   USBECK Laborgeräte
Clamps
Catalog # 241-7547
Clamps
Clamps
30 - 80 mm
Price
231,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Zinc die-casting, nickel plated.
  • With vinyl coated jaws
  • With rod Ø 10 mm
Specifications
  • Spændvidde:
    30 - 80 mm