Orders LinkContactUsLinkComponent
Dobbeltmuffer
With nickel plated brass screws, Zinc die-cast, powder coated, with nickel plated brass screws, 16,0 mm
Katalog # 241-7222
Leverandør:   USBECK Laborgeräte
Dobbeltmuffer
Katalog # 241-7222
Leverandør:  USBECK Laborgeräte
Dobbeltmuffer
Dobbeltmuffer
With nickel plated brass screws, Zinc die-cast, powder coated, with nickel plated brass screws, 16,0 mm
Price
171,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Til montering på stativ
  • Hurtig og let at samle og skille ad
  • Robust, holdbar
  • Forskellige materialer til mange applikationer
Specifikationer
  • Beskrivelse:
    With nickel plated brass screws
  • Materiale:
    Zinc die-cast, powder coated, with nickel plated brass screws
  • Seq:
    7
  • Til stang Ø:
    16,0 mm