Evaporating dishes
75 ml, 20 mm, 101 mm
Katalog # 216-0294
Leverandør: TECHNICAL QUARTZ SOLUTIONS
Gå til:
Product Details & Documents
Made of silica.
- Very good chemical resistance
- Very good temperature resistance
- Easy to clean
Specifikationer
- Udv.Ø:101 mm
- Kapacitet:75 ml
- Højde:20 mm