Orders LinkContactUsLinkComponent
Aluminium headspace caps
Aluminium headspace caps
Aluminium cap, star spring and septa, Butyl red
Katalog # PERCB0159356
Leverandør:   PerkinElmer
Aluminium headspace caps
Katalog # PERCB0159356
Leverandør:  PerkinElmer
Aluminium headspace caps
Aluminium cap, star spring and septa, Butyl red
Price
1.100,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Un-Assembled aluminum caps and septa for all of your analytical and biological applications.
  • Temperature resistance up to 130 °C
  • Very economical and has low resistance to coring
Specifikationer
  • Septum:
    Butyl red
  • Beskrivelse:
    Aluminium cap, star spring and septa
  • Seq:
    04