Aluminium headspace caps
Aluminium cap, star spring and septa, Butyl red
Katalog # PERCB0159356
Leverandør: PerkinElmer
Gå til:
Product Details & Documents
Un-Assembled aluminum caps and septa for all of your analytical and biological applications.
- Temperature resistance up to 130 °C
- Very economical and has low resistance to coring
Specifikationer
- Septum:Butyl red
- Beskrivelse:Aluminium cap, star spring and septa
- Seq:04