Latex tests, Dryspot
Legionella Pneumophilia Serogroup 1, 50 Tests
Katalog # OXOIDR0801M
Leverandør: THERMO FISHER DIAGNOSTICS
Gå til:
Product Details & Documents
Dry latex test designed to demonstrate by slide agglutination strains possessing a serogroup antigen.
- Simplified room temperature storage allows storage for up to two years
- Blue latex particles are sensitized with reactive (positive) and non-reactive (negative) antibodies and dried onto reaction cards
Certifikater
Søg efter partinummerSpecifikationer
Specifikationer
- Pakn:50 Tests
- Beskrivelse:Legionella Pneumophilia Serogroup 1