Orders LinkContactUsLinkComponent
Latex tests, Dryspot
Latex tests, Dryspot
Legionella Testkit, 50 Tests
  OXOIDR0800M
Latex tests, Dryspot
  OXOIDR0800M
 :  
Latex tests, Dryspot
Legionella Testkit, 50 Tests
 :  
7.300,00 kr
1 * 1 Each
Dry latex test designed to demonstrate by slide agglutination strains possessing a serogroup antigen.
  • Simplified room temperature storage allows storage for up to two years
  • Blue latex particles are sensitized with reactive (positive) and non-reactive (negative) antibodies and dried onto reaction cards
Documents
  • Pakn:
    50 Tests
  • Beskrivelse:
    Legionella Testkit