Orders LinkContactUsLinkComponent
Latex tests, Dryspot
Sb Only Streptococ.Extract.Enzyme 2X12Ml, 2
  OXOIDR0593G
Latex tests, Dryspot
  OXOIDR0593G
 :  
Latex tests, Dryspot
Sb Only Streptococ.Extract.Enzyme 2X12Ml, 2
 :  
676,00 kr
1 * 2 Items
Dry latex test designed to demonstrate by slide agglutination strains possessing a serogroup antigen.
  • Simplified room temperature storage allows storage for up to two years
  • Blue latex particles are sensitized with reactive (positive) and non-reactive (negative) antibodies and dried onto reaction cards
  • Pakn:
    2
  • Beskrivelse:
    Sb Only Streptococ.Extract.Enzyme 2X12Ml