Latex tests, Dryspot
Sb Only Streptococ.Extract.Enzyme 2X12Ml, 2
OXOIDR0593G
Dry latex test designed to demonstrate by slide agglutination strains possessing a serogroup antigen.
- Simplified room temperature storage allows storage for up to two years
- Blue latex particles are sensitized with reactive (positive) and non-reactive (negative) antibodies and dried onto reaction cards
- Pakn:2
- Beskrivelse:Sb Only Streptococ.Extract.Enzyme 2X12Ml