Orders LinkContactUsLinkComponent
Latex tests, Dryspot
Latex tests, Dryspot
Dryspot Staphytect Plus kit, 120 Tests
Katalog # OXOIDR0100M
Latex tests, Dryspot
Katalog # OXOIDR0100M
CAS nummer:  
Latex tests, Dryspot
Dryspot Staphytect Plus kit, 120 Tests
CAS nummer:  
Price
1.900,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Dry latex test designed to demonstrate by slide agglutination strains possessing a serogroup antigen.
  • Simplified room temperature storage allows storage for up to two years
  • Blue latex particles are sensitized with reactive (positive) and non-reactive (negative) antibodies and dried onto reaction cards
Documents
Specifikationer
Specifikationer
  • Pakn:
    120 Tests
  • Beskrivelse:
    Dryspot Staphytect Plus kit