Labels for slides
Slide labels on sheets, Papir, Sort på hvid, 22×22 mm
630-1837
These thin labels are suitable for use with microscopy slides.
- Permanent adhesive
- No curling or detaching
Some models can be customised: Imprint can be on the top or bottom of the label
Farve: Hvid
- Farve:Sort på hvid
- Beskrivelse:Slide labels on sheets
- Dimensioner:22×22 mm
- Materiale:Papir
- Seq:003